Sulphanilamide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Aminobenzenesulphonamide
M-Clarity™ quality level = MQ200
S9251-100G S9251-500G
SIALS9251-100GEA 61.1 EUR
SIALS9251-100G SIALS9251-500G
Sulphanilamide, Sigma-Aldrich®
Sulphanilamide
Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: 401
Melting Pt: 163…167 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
EINECS: 200-563-4
Merck Index: 13,09010
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM