Sulphanilic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid
M-Clarity™ quality level = MQ200
251917-100G
251917-25G
251917-500G
SIAL251917-100GEA
100
EUR
SIAL251917-100G
SIAL251917-25G
SIAL251917-500G
Sulphanilic acid, Sigma-Aldrich®
Sulphanilic acid
Formula:
4-(H₂N)C₆H₄SO₃H MW: 173.19 g/mol Boiling Pt: 363 Melting Pt: 279 Density: 1.485 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007886 CAS Number: 121-57-3 EINECS: 204-482-5 Merck Index: 13,09012 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...