Sulphanilic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid
86090-100G 86090-500G 86090-5G
SIAL86090-100GEA 108 EUR
SIAL86090-100G SIAL86090-500G SIAL86090-5G
Sulphanilic acid, Sigma-Aldrich®
Sulphanilic acid
Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM