O-Phenylenediamine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD

M-Clarity™ quality level = MQ200

P23938-500G P23938-5G P23938-100G
SIALP23938-500GEA 64.9 EUR
SIALP23938-500G SIALP23938-5G SIALP23938-100G
O-Phenylenediamine, Sigma-Aldrich®
o-Phenylenediamine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 257
Melting Pt: 103
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00007721
CAS Number: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1 III
Merck Index: 12,07438

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR