O-Phenylenediamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD
M-Clarity™ quality level = MQ200
P5412-100TAB
P5412-50TAB
SIALP5412-100TABEA
989
EUR
SIALP5412-100TAB
SIALP5412-50TAB
O-Phenylenediamine, Sigma-Aldrich®
o-Phenylenediamine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 257 Melting Pt: 103 Density: 1.21 g/cm³ (20 °C) Flash Pt: 110 Storage Temperature: Ambient |
MDL Number:
MFCD00007721 CAS Number: 95-54-5 EINECS: 202-430-6 UN: 1673 ADR: 6.1 III Merck Index: 12,07438 |