O-Phenylenediamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD
M-Clarity™ quality level = MQ200
P9029-100G
P9029-50G
SIALP9029-100GEA
108
EUR
SIALP9029-100G
SIALP9029-50G
O-Phenylenediamine, Sigma-Aldrich®
o-Phenylenediamine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 257 °C (1013 hPa) Melting Pt: 103 °C Density: 1.21 g/cm³ (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007721 CAS Number: 95-54-5 EINECS: 202-430-6 UN: 1673 ADR: 6.1,III Merck Index: 12,07438 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...