m-Phenylenediamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: MPD, 1,3-Diaminobenzene, 1,3-Phenylenediamine
M-Clarity™ quality level = MQ200
P23954-100G P23954-15KG P23954-1KG P23954-25G P23954-500G P23954-5G
SIALP23954-100GEA 32.5 EUR
SIALP23954-100G SIALP23954-15KG SIALP23954-1KG SIALP23954-25G SIALP23954-500G SIALP23954-5G
m-Phenylenediamine, Sigma-Aldrich®
m-Phenylenediamine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 63…64 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 187 °C
Storage Temperature: Ambient
MDL Number: MFCD00007799
CAS Number: 108-45-2
EINECS: 203-584-7
UN: 1673
ADR: 6.1,III
Merck Index: 12,07437