p-Phenylenediamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,4-Diaminobenzene, PPD, 1,4-Phenylenediamine
M-Clarity™ quality level = MQ200
695106-1G 695106-5G
SIAL695106-1GEA 45.7 EUR
SIAL695106-1G SIAL695106-5G
p-Phenylenediamine, Sigma-Aldrich®
p-Phenylenediamine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 267
Melting Pt: 139…141 °C
Density: 1.251 g/cm³ (20 °C)
Flash Pt: 156
Storage Temperature: Ambient
MDL Number: MFCD00007901
CAS Number: 106-50-3
EINECS: 203-404-7
UN: 1673
ADR: 6.1 III
Merck Index: 12,07439