N-Methylaniline, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
236233-10G 236233-50G
SIAL236233-10GEA 115 EUR
SIAL236233-10G SIAL236233-50G
N-Methylaniline, Sigma-Aldrich®
N-Methylaniline
Formula: C₇H₉N
Boiling Pt: 196.3 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.989 g/cm³ (20 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008283
CAS Number: 100-61-8
EINECS: 202-870-9
UN: 2294
ADR: 6.1,III
Merck Index: 12,06098