Diphenylamine
Supplier: EHRENSTORFER GMBH
EHERC12890000EA
29.6
EUR
EHERC12890000
Diphenylamine
Diphenylamine
Formula:
(C₆H₅)₂NH MW: 169,23 g/mol Boiling Pt: 302 °C (1013 hPa) Melting Pt: 53…54 °C Density: 1,165 g/cm³ (20 °C) Flash Pt: 153 °C (closed cup) |
MDL Number:
MFCD00003014 UN: 3077 ADR: 9,III Merck Index: 13,03349 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...