Diphenylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
N-Phenylaniline, N-Phenylbenzeneamine
M-Clarity™ quality level = MQ100
112763-100G
112763-1KG
SIAL112763-100GEA
85.7
EUR
SIAL112763-100G
SIAL112763-1KG
Diphenylamine, Sigma-Aldrich®
Diphenylamine
Formula:
(C₆H₅)₂NH MW: 169.23 g/mol Boiling Pt: 302 °C (1013 hPa) Melting Pt: 53…54 °C Density: 1.165 g/cm³ (20 °C) Flash Pt: 153 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003014 CAS Number: 122-39-4 EINECS: 204-539-4 UN: 3077 ADR: 9,III Merck Index: 13,03349 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...