4-Nitroanisole, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

103543-25G 103543-5G
SIAL103543-25GEA 50.5 EUR
SIAL103543-25G SIAL103543-5G
4-Nitroanisole, Sigma-Aldrich®
4-Nitroanisole
Formula: C₇H₇NO₃
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 9.4 °C
Density: 1.254 g/cm³ (20 °C)
MDL Number: MFCD00007327
CAS Number: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1,III
Merck Index: 12,06682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR