4-Nitrobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Nitrobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ200
272248-100G 272248-25G 272248-5G
SIAL272248-100GEA 65.5 EUR
SIAL272248-100G SIAL272248-25G SIAL272248-5G
4-Nitrobenzenesulphonyl chloride, Sigma-Aldrich®
4-Nitrobenzenesulphonyl chloride
Formula: C₆H₄ClNO₄S
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007445
CAS Number: 98-74-8
EINECS: 202-697-9
UN: 3261
ADR: 8,II