Phenetole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL241989-100MLEA 46.2 EUR
SIAL241989-100ML SIAL241989-500ML
Phenetole, Sigma-Aldrich®
Phenetole
Formula: C₆H₅OC₂H₅
MW: 122,17 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0,956 g/cm³ (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00009090
UN: 3271
ADR: 3,III
Merck Index: 12,07374