Anisole, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

SIALW209708-SAMPLEEA 54.3 EUR
SIALW209708-SAMPLE SIALW209708-10KG-K SIALW209708-1KG-K
Anisole, Sigma-Aldrich®
Anisole
Formula: C₇H₈O
MW: 108,14 g/mol
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: –37 °C
Density: 0,9961 g/cm³ (20 °C)
Flash Pt: 44 °C
MDL Number: MFCD00008354
UN: 2222
ADR: 3,III
Merck Index: 13,00672
https://si.vwr.com/stibo/web/std.lang.all/96/93/759693.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR