2-Ethoxyphenol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Hydroxy-2-ethoxybenzene, Pyrocatechol monoethyl ether, o-Ethoxyphenol
M-Clarity™ quality level = MQ200
250910-100G
250910-500G
SIAL250910-100GEA
42.9
EUR
SIAL250910-100G
SIAL250910-500G
2-Ethoxyphenol, Sigma-Aldrich®
2-Ethoxyphenol
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 214…216 °C (1013 hPa) Melting Pt: 28…29 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002187 CAS Number: 94-71-3 EINECS: 202-358-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...