4-Ethoxyphenol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
258598-25G
258598-5G
SIAL258598-25GEA
79.2
EUR
SIAL258598-25G
SIAL258598-5G
4-Ethoxyphenol, Sigma-Aldrich®
4-Ethoxyphenol
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 246 Melting Pt: 63…64 °C |
MDL Number:
MFCD00002334 CAS Number: 622-62-8 EINECS: 210-748-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...