Guaiacol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, 2-Methoxyphenol
M-Clarity™ quality level = MQ100
W253200-10KG-K W253200-1KG-K W253200-5KG-K
SIALW253200-10KG-KEA 765 EUR
SIALW253200-10KG-K SIALW253200-1KG-K SIALW253200-5KG-K
Guaiacol, Sigma-Aldrich®
Guaiacol
Formula: C₇H₈O₂
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1.1534 g/cm³ (0 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
UN: 2810
ADR: 6.1,III
Merck Index: 13,04568