3-Methoxyphenol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Hydroxy-3-methoxybenzene, m-Methoxyphenol

M-Clarity™ quality level = MQ100

328456-100G 328456-25G
SIAL328456-100GEA 78.6 EUR
SIAL328456-100G SIAL328456-25G
3-Methoxyphenol, Sigma-Aldrich®
3-Methoxyphenol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.131 g/cm³ (20 °C)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00002267
CAS Number: 150-19-6
EINECS: 205-754-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR