2-Methoxythiophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W415901-100G W415901-1KG W415901-25G W415901-500G W415901-SAMPLE
SIALW415901-100GEA 256 EUR
SIALW415901-100G SIALW415901-1KG SIALW415901-25G SIALW415901-500G SIALW415901-SAMPLE
2-Methoxythiophenol, Sigma-Aldrich®
2-Methoxythiophenol
Formula: C₇H₈OS
Boiling Pt: 225 °C (1013 hPa)
Density: 1.14 g/cm³ (30 °C)
MDL Number: MFCD00004834
CAS Number: 7217-59-6
EINECS: 230-605-7
UN: 3334
ADR: 9,