3-Methoxythiophenol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-Methoxybenzenethiol, 1-Methoxy-3-mercaptobenzene, 3-Mercaptoanisol, 3-Methoxybenzene-1-thiol, 3-Methoxyphenyl mercaptan, m-Methoxythiophenol
155705-25G
155705-5G
SIAL155705-25GEA
134
EUR
SIAL155705-25G
SIAL155705-5G
3-Methoxythiophenol, Sigma-Aldrich®
3-Methoxythiophenol
Formula:
C₇H₈OS Boiling Pt: 223 °C (1013 hPa) Density: 1.136 g/cm³ (20 °C) |
MDL Number:
MFCD00004841 CAS Number: 15570-12-4 EINECS: 239-617-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...