1,3-Dimethoxybenzene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Resorcinol dimethyl ether
M-Clarity™ quality level = MQ200
126306-100G 126306-500G
SIAL126306-100GEA 33.1 EUR
SIAL126306-100G SIAL126306-500G
1,3-Dimethoxybenzene, Sigma-Aldrich®
1,3-Dimethoxybenzene
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008384
CAS Number: 151-10-0
EINECS: 205-783-4