1,4-Dimethoxybenzene, Sigma-Aldrich®
Supplier: Merck
Synonyms: p-Dimethoxybenzene, Hydroquinone dimethyl ether
M-Clarity™ quality level = MQ300
W238600-10KG-K W238600-1KG-K W238600-5KG-K
SIALW238600-10KG-KEA 617 EUR
SIALW238600-10KG-K SIALW238600-1KG-K SIALW238600-5KG-K
1,4-Dimethoxybenzene, Sigma-Aldrich®
1,4-Dimethoxybenzene
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 212.6 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 0.874 g/cm³ (24.9 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008401
CAS Number: 150-78-7
EINECS: 205-771-9