3,5-Dimethoxyphenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
132632-10G 132632-50G
SIAL132632-10GEA 88.7 EUR
SIAL132632-10G SIAL132632-50G
3,5-Dimethoxyphenol, Sigma-Aldrich®
3,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 294
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00008388
CAS Number: 500-99-2
EINECS: 207-917-7