Resorcinol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1,3-Dihydroxybenzene, 1,3-Benzenediol
MQ100
R5645-100G
R5645-500G
SIALR5645-100GEA
109
EUR
SIALR5645-100G
SIALR5645-500G
Resorcinol, Sigma-Aldrich®
Resorcinol
Formula:
C₆H₆O₂ MW: 110.11 g/mol Boiling Pt: 281 Melting Pt: 109…111 °C Density: 1.272 g/cm³ (20 °C) Flash Pt: 127 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002269 CAS Number: 108-46-3 EINECS: 203-585-2 UN: 2876 ADR: 6.1 III Merck Index: 12,08323 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...