Resorcinol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,3-Dihydroxybenzene, 1,3-Benzenediol
MQ300
W358908-10KG W358908-1KG W358908-5KG W358908-SAMPLE
SIALW358908-10KGEA 490 EUR
SIALW358908-10KG SIALW358908-1KG SIALW358908-5KG SIALW358908-SAMPLE
Resorcinol, Sigma-Aldrich®
Resorcinol
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 281
Melting Pt: 109…111 °C
Density: 1.272 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002269
CAS Number: 108-46-3
EINECS: 203-585-2
UN: 2876
ADR: 6.1 III
Merck Index: 12,08323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fa2900537bdb42ccfd55e4318288eb881a61a0&vid=e62c617319ff4711a7ff00e69774ad92cfd75d09&prid=web&clid=SAPDAM