Hydroquinone, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,4-Dihydroxybenzene, Quinol
M-Clarity™ quality level = MQ200
H17902-100G
H17902-2KG
H17902-500G
SIALH17902-100GEA
34
EUR
SIALH17902-100G
SIALH17902-2KG
SIALH17902-500G
Hydroquinone, Sigma-Aldrich®
Hydroquinone
Formula:
C₆H₄(OH)₂ MW: 110.11 g/mol Boiling Pt: 285 °C (1013 hPa) Melting Pt: 172 °C Density: 1.332 g/cm³ (20 °C) Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9,III Merck Index: 13,04833 |