Hydroquinone, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,4-Dihydroxybenzene, Quinol
H9003-100G
H9003-500G
H9003-1KG
SIALH9003-100GEA
48.6
EUR
SIALH9003-100G
SIALH9003-500G
SIALH9003-1KG
Hydroquinone, Sigma-Aldrich®
Hydroquinone
Formula:
C₆H₄(OH)₂ MW: 110.11 g/mol Boiling Pt: 285 °C (1013 hPa) Melting Pt: 172 °C Density: 1.332 g/cm³ (20 °C) Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9,III Merck Index: 13,04833 |