Phloroglucinol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,3,5-Trihydroxybenzene
79330-100G 79330-25G
SIAL79330-100GEA 110 EUR
SIAL79330-100G SIAL79330-25G
Phloroglucinol, Sigma-Aldrich®
Phloroglucinol
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 400
Melting Pt: 217…220 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002286
CAS Number: 108-73-6
EINECS: 203-611-2
Merck Index: 12,07482