Diphenyl ether, Sigma-Aldrich®
Supplier: Merck
|
|
Synonyms:
Diphenyl oxide, Phenyl ether, Phenoxybenzene
P24101-1KG
P24101-3KG
SIALP24101-1KGEA
60.2
EUR
SIALP24101-1KG
SIALP24101-3KG
Diphenyl ether, Sigma-Aldrich®
Diphenyl ether
Formula:
C₁₂H₁₀O MW: 170.21 g/mol Boiling Pt: 498 °C (1013 hPa) Melting Pt: 24…27 °C Density: 1.07 g/cm³ (20 °C) Flash Pt: 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003034 CAS Number: 101-84-8 EINECS: 202-981-2 UN: 3077 ADR: 9,III Merck Index: 12,07442 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...