Diphenyl ether, Sigma-Aldrich®

Supplier: Merck

Synonyms: Diphenyl oxide, Phenyl ether, Phenoxybenzene

M-Clarity™ quality level = MQ300

W366706-10KG-K W366706-1KG-K W366706-25KG-K
SIALW366706-10KG-KEA 387 EUR
SIALW366706-10KG-K SIALW366706-1KG-K SIALW366706-25KG-K
Diphenyl ether, Sigma-Aldrich®
Diphenyl ether
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 498 °C (1013 hPa)
Melting Pt: 24…27 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS Number: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: 9,III
Merck Index: 12,07442

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR