Thiophenol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Mercaptobenzene, Phenyl mercaptan, Benzenethiol
M-Clarity™ quality level = MQ200
240249-10G 240249-50G
SIAL240249-10GEA 45.4 EUR
SIAL240249-10G SIAL240249-50G
Thiophenol, Sigma-Aldrich®
Thiophenol
Formula: C₆H₆S
MW: 110.18 g/mol
Boiling Pt: 169
Melting Pt: –15 °C
Density: 1.0728 g/cm³ (20 °C)
Flash Pt: 50
Storage Temperature: Ambient
MDL Number: MFCD00004826
CAS Number: 108-98-5
EINECS: 203-635-3
UN: 2337
ADR: 6.1 I
Merck Index: 12,09492