Benzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Benzene sulphochloride

M-Clarity™ quality level = MQ100

108138-100G 108138-1KG 108138-500G 108138-5G
SIAL108138-100GEA 37.8 EUR
SIAL108138-100G SIAL108138-1KG SIAL108138-500G SIAL108138-5G
Benzenesulphonyl chloride, Sigma-Aldrich®
Benzenesulphonyl chloride
Formula: C₆H₅ClO₂S
MW: 176.62 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.3842 g/cm³ (20 °C)
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8,III
Merck Index: 13,01072

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR