Benzenesulphonic acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
135070-1KG
135070-100G
SIAL135070-1KGEA
69.9
EUR
SIAL135070-1KG
SIAL135070-100G
Benzenesulphonic acid, Sigma-Aldrich®
Benzenesulphonic acid
Formula:
C₆H₆O₃S MW: 158.18 g/mol Boiling Pt: 319 °C (1013 hPa) Melting Pt: 50 °C Density: 1.32 g/cm³ (47 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011689 CAS Number: 98-11-3 EINECS: 202-638-7 UN: 2585 ADR: 8,III Merck Index: 13,01070 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...