4-Allylanisole, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Allyl-4-methoxybenzene, Estragole

M-Clarity™ quality level = MQ100

A29208-100G A29208-25G
SIALA29208-100GEA 80.5 EUR
SIALA29208-100G SIALA29208-25G
4-Allylanisole, Sigma-Aldrich®
4-Allylanisole
Formula: C₁₀H₁₂O
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 22.5 °C
Density: 0.965 g/cm³ (20 °C)
MDL Number: MFCD00008653
CAS Number: 140-67-0
EINECS: 205-427-8
Merck Index: 13,03740

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR