4-Allylanisole, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Allyl-4-methoxybenzene, Estragole
M-Clarity™ quality level = MQ300
W241105-5KG-K
W241105-1KG-K
SIALW241105-5KG-KEA
778
EUR
SIALW241105-5KG-K
SIALW241105-1KG-K
4-Allylanisole, Sigma-Aldrich®
4-Allylanisole
Formula:
C₁₀H₁₂O Boiling Pt: 216 °C (1013 hPa) Melting Pt: 22.5 °C Density: 0.965 g/cm³ (20 °C) |
MDL Number:
MFCD00008653 CAS Number: 140-67-0 EINECS: 205-427-8 Merck Index: 13,03740 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...