Eugenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
M-Clarity™ quality level = MQ300
W246700-1KG-K W246700-5KG-K
SIALW246700-1KG-KEA 121 EUR
SIALW246700-1KG-K SIALW246700-5KG-K
Eugenol, Sigma-Aldrich®
Eugenol
Formula: C₁₀H₁₂O₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: ∼–10 °C
Density: 1.0652 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931