α-Methylstyrene, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2-Phenylpropene, Isopropenylbenzene

M-Clarity™ quality level = MQ200

M80903-5ML M80903-100ML M80903-1L M80903-2.5L
SIALM80903-5MLEA 26.7 EUR
SIALM80903-5ML SIALM80903-100ML SIALM80903-1L SIALM80903-2.5L
α-Methylstyrene, Sigma-Aldrich®
α-Methylstyrene
Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 165.4 °C (1013 hPa)
Melting Pt: –23 °C
Density: 0.9108 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008859
CAS Number: 98-83-9
EINECS: 202-705-0
UN: 2303
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR