Styrene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phenylethene, Phenylethylene, Vinylbenzene
MQ200
S4972-1L S4972-500ML S4972-5ML
SIALS4972-1LEA 51.1 EUR
SIALS4972-1L SIALS4972-500ML SIALS4972-5ML
Styrene, Sigma-Aldrich®
Styrene
Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 145.2
Melting Pt: –31 °C
Density: 0.906 g/cm³ (20 °C)
Flash Pt: 31
MDL Number: MFCD00008612
CAS Number: 100-42-5
EINECS: 202-851-5
UN: 2055
ADR: 3 III
Merck Index: 13,08944