trans-Chalcone, Sigma-Aldrich®
Supplier: Merck
Synonyms: trans-Benzylidenacetophenone, (E)-Chalcone, trans-Benzalacetophenone, (E)-1,3-Diphenylprop-2-en-1-one
M-Clarity™ quality level = MQ100
136123-100G 136123-5G
SIAL136123-100GEA 53.3 EUR
SIAL136123-100G SIAL136123-5G
trans-Chalcone, Sigma-Aldrich®
trans-Chalcone
Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 347
Melting Pt: 55…57 °C
MDL Number: MFCD00003082
CAS Number: 614-47-1
EINECS: 210-383-8
Merck Index: 13,02028
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f708bafe7d3c2e194d3e9bf5f410a2c30afb756&vid=aa90a6b415f359c4551772765e81cbdff2cce599&prid=web&clid=SAPDAM