trans-Stilbene, Sigma-Aldrich®
Supplier: Merck
Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene
M-Clarity™ quality level = MQ100
139939-100G 139939-25G
SIAL139939-100GEA 173 EUR
SIAL139939-100G SIAL139939-25G
trans-Stilbene, Sigma-Aldrich®
trans-Stilbene
Formula: C₁₄H₁₂
MW: 180.25 g/mol
Boiling Pt: 306
Melting Pt: 121…124 °C
Density: 0.9707 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064300
CAS Number: 103-30-0
EINECS: 203-098-5
Merck Index: 12,08972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52acf17325a810eecda8f5a4d795a5b31a1f6434&vid=6d95185bf3c5a4ec46f2df78329eb0b669b2b255&prid=web&clid=SAPDAM