1-Chloro-4-ethynylbenzene, Sigma-Aldrich®

Supplier: Merck

SIAL206474-1GEA 63.6 EUR
SIAL206474-1G
1-Chloro-4-ethynylbenzene, Sigma-Aldrich®
1-Chloro-4-ethynylbenzene
Formula: C₈H₅Cl
MW: 136,58 g/mol
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR