Phenylacetylene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
117706-25ML
117706-100ML
SIAL117706-25MLEA
46.2
EUR
SIAL117706-25ML
SIAL117706-100ML
Phenylacetylene, Sigma-Aldrich®
Phenylacetylene
Formula:
C₈H₆ MW: 102.14 g/mol Boiling Pt: 144 °C (1013 hPa) Melting Pt: –44.8 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 31 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008570 CAS Number: 536-74-3 EINECS: 208-645-1 UN: 3295 ADR: 3,III Merck Index: 13,03892 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...