Diphenylacetylene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

D204803-25G D204803-5G
SIALD204803-25GEA 215 EUR
SIALD204803-25G SIALD204803-5G
Diphenylacetylene, Sigma-Aldrich®
Diphenylacetylene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 56…59 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004786
CAS Number: 501-65-5
EINECS: 207-926-6
Merck Index: 12,09643

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR