(3-Bromopropyl)benzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Bromo-3-phenylpropane
M-Clarity™ quality level = MQ200
B77207-100G B77207-5G
SIALB77207-100GEA 197 EUR
SIALB77207-100G SIALB77207-5G
(3-Bromopropyl)benzene, Sigma-Aldrich®
(3-Bromopropyl)benzene
Formula: BrH₂CCH₂CH₂(C₆H₅)
MW: 199.09 g/mol
Boiling Pt: 219 °C (1013 hPa)
Density: 1.3003 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00000257
CAS Number: 637-59-2
EINECS: 211-294-7