Benzyl bromide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: alpha-Bromotoluene
MQ200
B17905-100G B17905-25G B17905-500G
SIALB17905-100GEA 52.3 EUR
SIALB17905-100G SIALB17905-25G SIALB17905-500G
Benzyl bromide, Sigma-Aldrich®
Benzyl bromide
Formula: (C₆H₅)CH₂Br
MW: 171.04 g/mol
Boiling Pt: 199
Melting Pt: –4 °C
Density: 1.438 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00000172
CAS Number: 100-39-0
EINECS: 202-847-3
UN: 1737
ADR: 6.1 II
Merck Index: 13,01130