α,3-Dibromotoluene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene
187062-25G 187062-5G
SIAL187062-25GEA 51.2 EUR
SIAL187062-25G SIAL187062-5G
α,3-Dibromotoluene, Sigma-Aldrich®
α,3-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 258
Melting Pt: 41…44 °C
Density: 1.56 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000176
CAS Number: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8 II