1-Bromo-4-butylbenzene 97%
Supplier: Thermo Scientific
Warning
Synonyms: (4-Bromophenyl)butane
320440100 320440500
ACRO320440100EA 30.1 EUR
ACRO320440100 ACRO320440500
1-Bromo-4-butylbenzene 97%
1-Bromo-4-butylbenzene
Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: -26 °C
Density: 1.2 g/cm³
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00040934
CAS Number: 41492-05-1
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5330 (20°C, 589 nm)
GC ≥96.0 %