1-Bromo-4-pentylbenzene

Supplier: Thermo Scientific

BTB09867.10 BTB09867.50
MAYBBTB09867.10EA 0 EUR
MAYBBTB09867.10 MAYBBTB09867.50
1-Bromo-4-pentylbenzene
1-Bromo-4-pentylbenzene
Formula: C₁₁H₁₅Br
MDL Number: MFCD00061113
CAS Number: 51554-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR