α-Bromo-2,5-dichlorotoluene, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2,5-Dichlorobenzyl bromide, 2-(Bromomethyl)-1,4-dichlorobenzene

M-Clarity™ quality level = MQ100

548987-25G 548987-5G
SIAL548987-25GEA 182 EUR
SIAL548987-25G SIAL548987-5G
α-Bromo-2,5-dichlorotoluene, Sigma-Aldrich®
α-Bromo-2,5-dichlorotoluene
Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 39…41 °C
MDL Number: MFCD00236034
CAS Number: 85482-13-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR